Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Positive test controls include: 293T.
Immunogen sequence: NNELSPVALR QMSCAAGTTQ TACTDDNALA YYNTTKGGRF VLALLSDLQD LKWARFPKSD GTGTIYTELE PPCRFVTDTP KGPKVKYLYF IKGLNNLNRG MVLGSLAATV RLQ
Severe Acute Respiratory Syndrome (SARS), an emerging disease characterized by atypical pneumonia, has recently been attributed to a novel coronavirus (SARS-CoV). The nucleocapsid protein (N) was one of the structural proteins of SARS-CoV (N, S, sE, M). The severe acute respiratory syndrome coronavirus (SARS-CoV) nucleocapsid (N) protein is the most abundant protein in the virus-infected cells. The SARS-CoV-2 S gene has 76% amino acid similarity to the SARS-CoV S gene, and nonsynonymous mutations developed in the S protein as the SARS-CoV epidemic developed, reports have shown that the crystal structure of the SARS-CoV-2 nucleocapsid protein is similar to those of previously described coronavirus N proteins, but their surface electrostatic potential characteristics are distinct.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CoV; CoV-2; NSP9; SARS
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support