Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: KECPLSQSMI SSIVNSTYYA NVSATKCQEF GRWYKKYKKI KVERVERENL SDYCVLGQRP MHLPNMNQLA SLGKTNEQSP HSQIHHSTPI RNQVPALQPI MSPGLLSPQL SPQLV
DNA-binding protein SATB2 binds to DNA at the nuclear matrix- or scaffold associated regions. It is thought to recognize the sugar-phosphate structure of double-stranded DNA. SATB2 is a transcription factor controlling nuclear gene expression by binding to matrix attachment regions (MARs) of DNA and inducing a local chromatin-loop remodeling. It also acts as a docking site for several chromatin remodeling enzymes by recruiting corepressors (HDACs) or coactivators (HATs) directly to promoters and enhancers. SATB2 is required for the initiation of the upper-layer neurons (UL1) specific genetic program and for the inactivation of deep-layer neurons (DL) and UL2 specific genes - probably by modulating BCL11B expression. It is a repressor of Ctip2 and a regulatory determinant of corticocortical connections in the developing cerebral cortex. Mutations in the gene are associate with cleft palate isolated (CPI).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DNA-binding protein SATB2; FLJ21474; FLJ32076; MGC119474; MGC119477; SATB family member 2; special AT-rich sequence binding protein 2; Special AT-rich sequence-binding protein 2
Gene Aliases: GLSS; KIAA1034; RGD1562369; SATB2
UniProt ID: (Human) Q9UPW6
Entrez Gene ID: (Human) 23314, (Rat) 501145
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support