Search Thermo Fisher Scientific
FIGURE: 1 / 2
Description: This SNF 2C4 monoclonal antibody recognizes human SMARCA1 protein.
Applications Reported: This SNF 2C4 antibody has been reported for use in western blot and immunocytochemistry (ICC).
Applications Tested: This SNF 2C4 antibody has been tested by western blot and ICC on cell lines. This may be used at less than or equal to 2.0 µg/mL for western blot and less than or equal to 5.0 µg/mL for ICC.
Members of this G protein family are characterized by the presence of conserved SNF2 domain, a helicase-like region. They also have an ATPase activity and are thought to have a significant role regulating chromatin remodeling. Although SNF2L1 has a ubiquitous expression pattern, its active form is expressed in neurons suggesting a possible role in promoting neurite outgrowth and regulating neuronal physiology. Principal Names: SWI/SNF related matrix associated actin dependent regulator of chromatin subfamily A member 1 (SMARCA1); SNF2L1; SNF2LB; Possible global transcription activator SNF2L1; SWI2 Official Gene Symbol: SMARCA1 Gen Bank Accession Number: NP_620604 Gene ID: 6594 (Human) Gene Map Locus: Xq25 (Human) SNF2L1, a ~123KDa protein, is a member of SWI/SNF family of proteins.