Search
Search
OriGene
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Catalyzes the production of spermine from spermidine and decarboxylated S-adenosylmethionine (dcSAM).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Spermidine aminopropyltransferase; Spermine synthase; SPMSY; unnamed protein product
Gene Aliases: AI427066; Gy; gyro; MRSR; MRXSSR; SMS; SpmST; SPMSY; SpS; SRS
UniProt ID: (Human) P52788, (Mouse) P97355
Entrez Gene ID: (Dog) 480861, (Human) 6611, (Mouse) 20603, (Rat) 363469
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support