Search Thermo Fisher Scientific
FIGURE: 1 / 4
Recommended positive controls: A549, H1299, HCT116, H1299 nuclear extract.
Predicted reactivity: Mouse (95%), Rat (96%), Chicken (89%), Rhesus Monkey (99%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
SNRK may play a role in hematopoietic cell proliferation or differentiation. SNRK is the potential mediator of neuronal apoptosis.