Search Thermo Fisher Scientific
A 1:1,000 dilution of MA5-45917 was sufficient for detection of SPATA2L in 10 µg of SH-SY5Y Cell Line by ECL immunoblot analysis using goat anti-mouse IgG:HRP as the secondary antibody.|Detects 50 kDa.
SPATA2L belongs to the SPATA2 family. The exact function of SPATA2L is not known. There are two different isoforms.