Product References
Nuclear receptor corepressors Ncor1 and Ncor2 (Smrt) are required for retinoic acid-dependent repression of Fgf8 during somitogenesis.
Developmental biology
Kumar S,Cunningham TJ,Duester G
MA1840 was used in ChIP assay to propose a model in which NCOR1/2 mediates retinoic acid-dependent repression of Fgf8 in caudal progenitors
Sat Oct 01 00:00:00 EDT 2016
SIP1/NHERF2 enhances estrogen receptor alpha transactivation in breast cancer cells.
Nucleic acids research
Meneses-Morales I,Tecalco-Cruz AC,Barrios-García T,Gómez-Romero V,Trujillo-González I,Reyes-Carmona S,García-Zepeda E,Méndez-Enríquez E,Cervantes-Roldán R,Pérez-Sánchez V,Recillas-Targa F,Mohar-Betancourt A,León-Del-Río A
MA1-840 was used in immunoprecipitation and western blot to study the role of NHERF2 as an ER-alpha co-activator in breast cancer cells
Sun Jun 01 00:00:00 EDT 2014
SIP1/NHERF2 enhances estrogen receptor alpha transactivation in breast cancer cells.
Nucleic acids research
Meneses-Morales I,Tecalco-Cruz AC,Barrios-García T,Gómez-Romero V,Trujillo-González I,Reyes-Carmona S,García-Zepeda E,Méndez-Enríquez E,Cervantes-Roldán R,Pérez-Sánchez V,Recillas-Targa F,Mohar-Betancourt A,León-Del-Río A
MA1-840 was used in immunoprecipitation and western blot to study the role of NHERF2 as an ER-alpha co-activator in breast cancer cells
Sun Jun 01 00:00:00 EDT 2014
D538G mutation in estrogen receptor-α: A novel mechanism for acquired endocrine resistance in breast cancer.
Cancer research
Merenbakh-Lamin K,Ben-Baruch N,Yeheskel A,Dvir A,Soussan-Gutman L,Jeselsohn R,Yelensky R,Brown M,Miller VA,Sarid D,Rizel S,Klein B,Rubinek T,Wolf I
MA1-840 was used in immunoprecipitation to study the role of ER-alpha D538G mutation in mediating aquired resistance to hormone therapy in breast cancer patients
Sun Dec 01 00:00:00 EST 2013
Expression patterns of the steroid receptor coactivator family in human ovarian endometriosis.
The journal of obstetrics and gynaecology research
Kumagami A,Ito A,Yoshida-Komiya H,Fujimori K,Sato A
MA1-840 was used in immunohistochemistry to characterize the expression of estrogen and steroid receptor cofactors in human endometriosis
Sat Oct 01 00:00:00 EDT 2011
Constitutive activation of the mitogen-activated protein kinase pathway impairs vitamin D signaling in human prostate epithelial cells.
Journal of cellular physiology
Zhang Z,Kovalenko P,Cui M,Desmet M,Clinton SK,Fleet JC
MA1-840 was used in western blot to investigate the effect of constitutive activation of MAPK signaling on vitamin D signaling in immortalized human prostate epithelial cells
Sun Aug 01 00:00:00 EDT 2010
Role of steroid receptor coactivators in glucocorticoid and transforming growth factor beta regulation of plasminogen activator inhibitor gene expression.
Molecular endocrinology (Baltimore, Md.)
Li G,Heaton JH,Gelehrter TD
MA1-840 was used in western blot to study the regulation of plasminogen activator inhibitor gene expression through steroid receptor coactivators.
Mon May 01 00:00:00 EDT 2006
Role of SRC-1 in the promotion of prostate cancer cell growth and tumor progression.
Cancer research
Agoulnik IU,Vaid A,Bingman WE,Erdeme H,Frolov A,Smith CL,Ayala G,Ittmann MM,Weigel NL
MA1-840 was used in immunohistochemistry to study the role of SRC-1 in prostate cancer cell growth and tumor progression.
Thu Sep 01 00:00:00 EDT 2005
Nuclear coactivator-62 kDa/Ski-interacting protein is a nuclear matrix-associated coactivator that may couple vitamin D receptor-mediated transcription and RNA splicing.
The Journal of biological chemistry
Zhang C,Dowd DR,Staal A,Gu C,Lian JB,van Wijnen AJ,Stein GS,MacDonald PN
MA1-840 was used in chromatin immunoprecipitation to investigate the role of nuclear coactivator-62 kDa/Ski-interacting protein (NCoA62/SKIP) during the vitamin D receptor (VDR)-mediated transcription and RNA splicing.
Fri Sep 12 00:00:00 EDT 2003
Ordered recruitment of histone acetyltransferases and the TRAP/Mediator complex to thyroid hormone-responsive promoters in vivo.
Proceedings of the National Academy of Sciences of the United States of America
Sharma D,Fondell JD
MA1-840 was used in western blot to study the role of histone acetylation in TRAP/Mediator complex recruitment and function.
Tue Jun 11 00:00:00 EDT 2002
Endogenously expressed estrogen receptor and coactivator AIB1 interact in MCF-7 human breast cancer cells.
Proceedings of the National Academy of Sciences of the United States of America
Tikkanen MK,Carter DJ,Harris AM,Le HM,Azorsa DO,Meltzer PS,Murdoch FE
MA1-840 was used in western blot to demonstrate the role of AIB1 in human estrogen receptor signal pathway in MCF-7 human breast cancer cells.
Tue Nov 07 00:00:00 EST 2000
Endogenously expressed estrogen receptor and coactivator AIB1 interact in MCF-7 human breast cancer cells.
Proceedings of the National Academy of Sciences of the United States of America
Tikkanen MK,Carter DJ,Harris AM,Le HM,Azorsa DO,Meltzer PS,Murdoch FE
MA1-840 was used in western blot to demonstrate the role of AIB1 in human estrogen receptor signal pathway in MCF-7 human breast cancer cells.
Tue Nov 07 00:00:00 EST 2000
Recruitment of nuclear receptor corepressor and coactivator to the retinoic acid receptor by retinoid ligands. Influence of DNA-heterodimer interactions.
The Journal of biological chemistry
Klein ES,Wang JW,Khalifa B,Gavigan SA,Chandraratna RA
MA1-840 was used in western blot to study the activation mechanism for retinoic acid receptors by their ligands.
Fri Jun 23 00:00:00 EDT 2000
Recruitment of nuclear receptor corepressor and coactivator to the retinoic acid receptor by retinoid ligands. Influence of DNA-heterodimer interactions.
The Journal of biological chemistry
Klein ES,Wang JW,Khalifa B,Gavigan SA,Chandraratna RA
MA1-840 was used in western blot to study the activation mechanism for retinoic acid receptors by their ligands.
Fri Jun 23 00:00:00 EDT 2000
Partial hormone resistance in mice with disruption of the steroid receptor coactivator-1 (SRC-1) gene.
Science (New York, N.Y.)
Xu J,Qiu Y,DeMayo FJ,Tsai SY,Tsai MJ,O'Malley BW
MA1-840 was used in western blot to investigate the functional properties of SRC-1 in mice
Fri Mar 20 00:00:00 EST 1998
Steroid receptor coactivator-1 is a histone acetyltransferase.
Nature
Spencer TE,Jenster G,Burcin MM,Allis CD,Zhou J,Mizzen CA,McKenna NJ,Onate SA,Tsai SY,Tsai MJ,O'Malley BW
MA1-840 was used in western blot to characterize the steroid receptor coactivator 1
Thu Sep 11 00:00:00 EDT 1997