Search Thermo Fisher Scientific
This antibody reacts with native and denatured large T-antigen. It does not react with the small T-antigen of SV40 or the large T-antigen of BKV, human papovavirus.
The SV40 T antigen is encoded by the early region of the SV40 genome. The large T antigen binds DNA, and complexes with a 53,000 dalton cellular protein, p53, which is required for initiation of viral DNA replication during lytic growth. In addition, the large T antigen binds DNA polymerase and the transcription factor AP-2 and forms a specific complex with the P105 product of the retinoblastoma susceptibility gene.