Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MAPITTSREE FDEIPTVVGI FSAFGLVFTV SLFAWICCQR KSSKSNKTPP YKFVHVLKGV DIYPENLNSK KKFGADDKNE VKNKPAVPKN SLHLDLEKR
May be involved in Ca2+-dependent exocytosis of secretory vesicles through Ca2+ and phospholipid binding to the C2 domain or may serve as Ca2+ sensors in the process of vesicular trafficking and exocytosis. Plays a role in dendrite formation by melanocytes (PubMed:23999003).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Synaptotagmin IV; Synaptotagmin-4; Synaptotagmin4; SytIV
Gene Aliases: HsT1192; KIAA1342; SYT4
UniProt ID: (Human) Q9H2B2
Entrez Gene ID: (Human) 6860
Molecular Function: membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support