Search Thermo Fisher Scientific
PA1-4934 detects Sua7p in budding yeast samples.
PA1-4934 has been successfully used in Western blot and ELISA procedures. By Western blot this antibody detects a ~41 kDa protein (Sua7p) in yeast samples.
PA1-4934 immunogen corresponds to the recombinant protein which was over-expressed in E. coli with a plasmid carrying the entire Sua7p protein (amino acid residues 1-346).
The fundamental transcription factor TFIIB has the characteristics of stabilizing the DNA binding of TATA box-binding protein (TBP) and binding directly to DNA by its conformational change. Also its N terminal region binds to the RNA channel of RNA polymerase undertaking a very important role in the determination of transcription initiation point and promoter clearance. Sua7p is the TFIIB of budding yeast and is composed of 346 amino acid residues.