Proteintech
Immunogen sequence: ELQLSVDCA NKTESDLSIE VEFEYPFRLH QVYFDAPTCR GGTTKVFLVG DYSSSAEFFV TVAVFAFLYS MGALATYIFL QNKYRENNKG PMLDFLATAV FAFMWLVSSS AWAKGLSDVK MATDPENIIK EMPVCRQTGN TCKELRDLVT SGLNTSVVFG FLNLVLWVGN LWFVFKETGW AAPFLRAPPG APEKQPAPGD AYGDAGYGQG PGGYGPQDSY GPQGGYQPDY GQPAGSGGSG YGPQGDYGQQ GYGPQGAPTS FSNQM (50-313 aa encoded by B C064550)
Synaptophysin is a calcium-binding and integral membrane glycoprotein present in presynaptic vesicles in almost all neurons. Synaptophysin has four transmembrane domains and it forms a complex with dynamin at high calcium concentrations suggesting an involvement in synaptic vesicle endocytosis. It is also involved in the regulation of short-term and long-term synaptic plasticity. Synaptophysin is currently the most widely used marker for nerve terminals and for differentiating neuroendocrine tumors. Mutations in the gene can result in mental retardation, X-linked 96.