Search Thermo Fisher Scientific
MA1-91859 detects TBP Associated Factor 1 from human samples. Cross reacts with human and weakly with mouse and rat.
MA1-91859 has been successfully used in Western blot applications. Positive control: HeLa nuclear lysate.
The MA1-91859 immunogen is: Full length protein (Human).
3-Phosphoglycerate dehydrogenase (PHGDH; EC 1.1.1.95) catalyzes the transition of 3-phosphoglycerate into 3-phosphohydroxypyruvate, which is the first and rate-limiting step in the phosphorylated pathway of serine biosynthesis, using NAD+/NADH as a cofactor.