Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TIPRL is an inhibitory regulator of protein phosphatase-2A.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CG9578-like; dJ69E11.3; MGC3794; OTTHUMP00000032581; OTTHUMP00000032582; Putative MAPK-activating protein PM10; TIP; TIP41, TOR signaling pathway regulator-like; TIP41-like protein; Type 2A-interacting protein
Gene Aliases: TIP; TIP41; TIPRL; TIPRL1
UniProt ID: (Human) O75663
Entrez Gene ID: (Human) 261726
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support