Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Potential channel-forming protein implicated in import of protein precursors into mitochondria.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Mitochondrial import receptor subunit TOM40B; Protein TOMM40-like; RP11-297K8.10; translocase of outer mitochondrial membrane 40 homolog (yeast)-like; translocase of outer mitochondrial membrane 40 homolog-like
Gene Aliases: TOMM40B; TOMM40L
UniProt ID: (Human) Q969M1
Entrez Gene ID: (Human) 84134
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support