Product References
Establishment of the iPSC line CUIMCi005-A from a patient with Stargardt disease for retinal organoid culture.
Stem cell research
Su PY,Lee W,Zernant J,Tsang SH,Nagasaki T,Corneo B,Allikmets R
MA1-023 was used in Immunocytochemistry-immunofluorescence to establish an induced pluripotent stem cell line for retinal organoid research from a patient with mild disease features who is compound heterozygous for the frequent c.5882G>A (p.Gly1961Glu) missense variant and a c.4947delC (p.Glu1650Argfs*12) frameshift variant.
Thu Dec 01 00:00:00 EST 2022
Simultaneous high-efficiency base editing and reprogramming of patient fibroblasts.
Stem cell reports
Jalil S,Keskinen T,Maldonado R,Sokka J,Trokovic R,Otonkoski T,Wartiovaara K
MA1-023 was used in Immunocytochemistry to show that overall, this method yields tens of gene-edited hiPSC monoclonal lines with unprecedented efficiency and robustness while considerably reducing the cell culture time and thus the risk for in vitro alterations.
Tue Dec 14 00:00:00 EST 2021
Simultaneous high-efficiency base editing and reprogramming of patient fibroblasts.
Stem cell reports
Jalil S,Keskinen T,Maldonado R,Sokka J,Trokovic R,Otonkoski T,Wartiovaara K
MA1-023 was used in Immunocytochemistry to show that overall, this method yields tens of gene-edited hiPSC monoclonal lines with unprecedented efficiency and robustness while considerably reducing the cell culture time and thus the risk for in vitro alterations.
Tue Dec 14 00:00:00 EST 2021
Generation of a human control iPS cell line (ESi080-A) from a donor with no rheumatic diseases.
Stem cell research
Castro-Viñuelas R,Sanjurjo-Rodríguez C,Piñeiro-Ramil M,Rodríguez-Fernández S,Fuentes-Boquete IM,Blanco FJ,Díaz-Prado SM
MA1-023 was used in Immunocytochemistry-immunoflourescence to report the establishment of the human iPS cell line N1-FiPS4F#7 generated from skin cells of a patient with no rheumatic diseases, thus obtaining an appropriate control iPS cell line for researchers working in the field of rheumatic diseases.
Sun Mar 01 00:00:00 EST 2020
Applying hydrodynamic pressure to efficiently generate induced pluripotent stem cells via reprogramming of centenarian skin fibroblasts.
PloS one
Vosough M,Ravaioli F,Zabulica M,Capri M,Garagnani P,Franceschi C,Piccand J,Kraus MR,Kannisto K,Gramignoli R,Strom SC
MA1-023 was used in Immunocytochemistry-immunoflourescence to show efficient generation of iPSCs from the elderly may provide a source of cells for the regeneration of tissues and organs with autologous cells as well as cellular models for the study of aging, longevity and age-related diseases.
Fri Feb 14 00:00:00 EST 2020
Functional changes of AMPA responses in human induced pluripotent stem cell-derived neural progenitors in fragile X syndrome.
Science signaling
Achuta VS,Möykkynen T,Peteri UK,Turconi G,Rivera C,Keinänen K,Castrén ML
MA1-023 was used in Immunocytochemistry-immunoflourescence to study the functional maturation of the excitatory transmission system in fragile X syndrome.
Tue Jan 16 00:00:00 EST 2018
Human pluripotent reprogramming with CRISPR activators.
Nature communications
Weltner J,Balboa D,Katayama S,Bespalov M,Krjutškov K,Jouhilahti EM,Trokovic R,Kere J,Otonkoski T
MA1-023 was used in Immunocytochemistry to demonstrate that human somatic cells can be programmed into induced pluripotent stem cells using only CRISPR-Cas9 based gene activation.
Fri Jul 06 00:00:00 EDT 2018
Human pluripotent reprogramming with CRISPR activators.
Nature communications
Weltner J,Balboa D,Katayama S,Bespalov M,Krjutškov K,Jouhilahti EM,Trokovic R,Kere J,Otonkoski T
MA1-023 was used in Immunocytochemistry to demonstrate that human somatic cells can be programmed into induced pluripotent stem cells using only CRISPR-Cas9 based gene activation.
Fri Jul 06 00:00:00 EDT 2018
Insulin mutations impair beta-cell development in a patient-derived iPSC model of neonatal diabetes.
eLife
Balboa D,Saarimäki-Vire J,Borshagovski D,Survila M,Lindholm P,Galli E,Eurola S,Ustinov J,Grym H,Huopio H,Partanen J,Wartiovaara K,Otonkoski T
MA1-023 was used in Immunocytochemistry to demonstrate that misfolded proinsulin impairs developing beta-cell proliferation without increasing apoptosis.
Fri Nov 09 00:00:00 EST 2018
ATPase-deficient mitochondrial inner membrane protein ATAD3A disturbs mitochondrial dynamics in dominant hereditary spastic paraplegia.
Human molecular genetics
Cooper HM,Yang Y,Ylikallio E,Khairullin R,Woldegebriel R,Lin KL,Euro L,Palin E,Wolf A,Trokovic R,Isohanni P,Kaakkola S,Auranen M,Lönnqvist T,Wanrooij S,Tyynismaa H
MA1-023 was used in Immunocytochemistry to show that mutations in ATPase family AAA-domain containing protein 3A can be dominantly inherited and underlie variable neurological phenotypes, including hereditary spastic paraplegia, with intrafamiliar variability.
Sat Apr 15 00:00:00 EDT 2017
Metabotropic glutamate receptor 5 responses dictate differentiation of neural progenitors to NMDA-responsive cells in fragile X syndrome.
Developmental neurobiology
Achuta VS,Grym H,Putkonen N,Louhivuori V,Kärkkäinen V,Koistinaho J,Roybon L,Castrén ML
MA1-023 was used in immunocytochemistry to study metabotropic glutamate receptor 5 in a murine model of fragile X syndrome
Sat Apr 01 00:00:00 EDT 2017
Amnion Epithelial Cells of Buffalo (Bubalus Bubalis) Term Placenta Expressed Embryonic Stem Cells Markers and Differentiated into Cells of Neurogenic Lineage In Vitro.
Animal biotechnology
Ghosh K,Selokar NL,Gahlawat SK,Kumar D,Kumar P,Yadav PS
MA1-023X was used in immunocytochemistry to isolate and characterize buffalo amniotic membrane-derived epithelial cells
Mon Jul 18 00:00:00 EDT 2016
Combined negative effect of donor age and time in culture on the reprogramming efficiency into induced pluripotent stem cells.
Stem cell research
Trokovic R,Weltner J,Noisa P,Raivio T,Otonkoski T
MA1-023 was used in immunocytochemistry to study the effects of donor age and passage number on the efficiency of iPSC induction
Wed Jul 01 00:00:00 EDT 2015
Generation of iPSC line HEL24.3 from human neonatal foreskin fibroblasts.
Stem cell research
Trokovic R,Weltner J,Otonkoski T
MA1-023 was used in immunocytochemistry to describe the creation of a human iPSC line HEL24.3.
Wed Jul 01 00:00:00 EDT 2015
Generation of iPSC line HEL47.2 from healthy human adult fibroblasts.
Stem cell research
Trokovic R,Weltner J,Otonkoski T
MA1-023 was used in immunocytochemistry to establish a human iPSC line
Wed Jul 01 00:00:00 EDT 2015