Product References
Perfluorooctanoic acid promotes pancreatic β cell dysfunction and apoptosis through ER stress and the ATF4/CHOP/TRIB3 pathway.
Environmental science and pollution research international
He X,Wu D,Xu Y,Zhang Y,Sun Y,Chang X,Zhu Y,Tang W
Thu Dec 01 00:00:00 EST 2022
Hsa_circRNA_0008028 Deficiency Ameliorates High Glucose-Induced Proliferation, Calcification, and Autophagy of Vascular Smooth Muscle Cells via miR-182-5p/TRIB3 Axis.
Oxidative medicine and cellular longevity
Shi L,Li Y,Shi M,Li X,Li G,Cen J,Liu D,Wei C,Lin Y
Wed Sep 07 00:00:00 EDT 2022
A systems biological analysis of the ATF4-GADD34-CHOP regulatory triangle upon endoplasmic reticulum stress.
FEBS open bio
Márton M,Bánhegyi G,Gyöngyösi N,Kálmán EÉ,Pettkó-Szandtner A,Káldi K,Kapuy O
Tue Nov 01 00:00:00 EDT 2022
Cannabidiol inhibits human glioma by induction of lethal mitophagy through activating TRPV4.
Autophagy
Huang T,Xu T,Wang Y,Zhou Y,Yu D,Wang Z,He L,Chen Z,Zhang Y,Davidson D,Dai Y,Hang C,Liu X,Yan C
Mon Nov 01 00:00:00 EDT 2021
p62/sequestosome 1 attenuates methylmercury-induced endoplasmic reticulum stress in mouse embryonic fibroblasts.
Toxicology letters
Takanezawa Y,Nakamura R,Sugimoto T,Ohshiro Y,Uraguchi S,Kiyono M
Wed Dec 15 00:00:00 EST 2021
TRIB3 regulates FSHR expression in human granulosa cells under high levels of free fatty acids.
Reproductive biology and endocrinology : RB&E
Wang N,Si C,Xia L,Wu X,Zhao S,Xu H,Ding Z,Niu Z
Thu Sep 09 00:00:00 EDT 2021
sTNFRII-Fc modification protects human UC-MSCs against apoptosis/autophagy induced by TNF-α and enhances their efficacy in alleviating inflammatory arthritis.
Stem cell research & therapy
Zhao Y,Yang X,Li S,Zhang B,Li S,Wang X,Wang Y,Jia C,Chang Y,Wei W
Sat Oct 09 00:00:00 EDT 2021
TRB3 mediates vascular remodeling by activating the MAPK signaling pathway in hypoxic pulmonary hypertension.
Respiratory research
Cao X,Fang X,Guo M,Li X,He Y,Xie M,Xu Y,Liu X
Tue Dec 14 00:00:00 EST 2021
Dietary saturated fatty acid palmitate promotes cartilage lesions and activates the unfolded protein response pathway in mouse knee joints.
PloS one
Tan L,Harper LR,Armstrong A,Carlson CS,Yammani RR
Mon Aug 23 00:00:00 EDT 2021
Age-Related Decline in Expression of Molecular Chaperones Induces Endoplasmic Reticulum Stress and Chondrocyte Apoptosis in Articular Cartilage.
Aging and disease
Tan L,Register TC,Yammani RR
Thu Oct 01 00:00:00 EDT 2020
High-fat diet induces endoplasmic reticulum stress to promote chondrocyte apoptosis in mouse knee joints.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Tan L,Harper L,McNulty MA,Carlson CS,Yammani RR
Wed Apr 01 00:00:00 EDT 2020
Proscillaridin A slows the prostate cancer progression through triggering the activation of endoplasmic reticulum stress.
Cell cycle (Georgetown, Tex.)
Wang F,Liu L,Tong Y,Li L,Liu Y,Gao WQ
Sun Mar 01 00:00:00 EST 2020
Down-regulation of TRIB3 inhibits the progression of ovarian cancer via MEK/ERK signaling pathway.
Cancer cell international
Wang S,Wang C,Li X,Hu Y,Gou R,Guo Q,Nie X,Liu J,Zhu L,Lin B
Sat Apr 16 00:00:00 EDT 2022
Down-regulation of TRIB3 inhibits the progression of ovarian cancer via MEK/ERK signaling pathway.
Cancer cell international
Wang S,Wang C,Li X,Hu Y,Gou R,Guo Q,Nie X,Liu J,Zhu L,Lin B
Sat Apr 16 00:00:00 EDT 2022
Sarcopenia is attenuated by TRB3 knockout in aging mice via the alleviation of atrophy and fibrosis of skeletal muscles.
Journal of cachexia, sarcopenia and muscle
Shang GK,Han L,Wang ZH,Liu YP,Yan SB,Sai WW,Wang D,Li YH,Zhang W,Zhong M
Sat Aug 01 00:00:00 EDT 2020
Sarcopenia is attenuated by TRB3 knockout in aging mice via the alleviation of atrophy and fibrosis of skeletal muscles.
Journal of cachexia, sarcopenia and muscle
Shang GK,Han L,Wang ZH,Liu YP,Yan SB,Sai WW,Wang D,Li YH,Zhang W,Zhong M
Sat Aug 01 00:00:00 EDT 2020
Design, synthesis, and biological evaluation of 5-((8-methoxy-2-methylquinolin-4-yl)amino)-1H-indole-2-carbohydrazide derivatives as novel Nur77 modulators.
European journal of medicinal chemistry
Li B,Yao J,Guo K,He F,Chen K,Lin Z,Liu S,Huang J,Wu Q,Fang M,Zeng J,Wu Z
Thu Oct 15 00:00:00 EDT 2020
Design, synthesis, and biological evaluation of 5-((8-methoxy-2-methylquinolin-4-yl)amino)-1H-indole-2-carbohydrazide derivatives as novel Nur77 modulators.
European journal of medicinal chemistry
Li B,Yao J,Guo K,He F,Chen K,Lin Z,Liu S,Huang J,Wu Q,Fang M,Zeng J,Wu Z
Thu Oct 15 00:00:00 EDT 2020
TRIB3 promotes hepatocellular carcinoma growth and predicts poor prognosis.
Cancer biomarkers : section A of Disease markers
Wang XJ,Li FF,Zhang YJ,Jiang M,Ren WH
Mon Jul 12 00:00:00 EDT 2021
TRIB3 promotes hepatocellular carcinoma growth and predicts poor prognosis.
Cancer biomarkers : section A of Disease markers
Wang XJ,Li FF,Zhang YJ,Jiang M,Ren WH
Mon Jul 12 00:00:00 EDT 2021
Wnt regulates amino acid transporter Slc7a5 and so constrains the integrated stress response in mouse embryos.
EMBO reports
Poncet N,Halley PA,Lipina C,Gierliński M,Dady A,Singer GA,Febrer M,Shi YB,Yamaguchi TP,Taylor PM,Storey KG
Tue Jan 07 00:00:00 EST 2020
Endoplasmic reticulum-targeting doxorubicin: a new tool effective against doxorubicin-resistant osteosarcoma.
Cellular and molecular life sciences : CMLS
Buondonno I,Gazzano E,Tavanti E,Chegaev K,Kopecka J,Fanelli M,Rolando B,Fruttero R,Gasco A,Hattinger C,Serra M,Riganti C
Fri Feb 01 00:00:00 EST 2019
Endoplasmic reticulum-targeting doxorubicin: a new tool effective against doxorubicin-resistant osteosarcoma.
Cellular and molecular life sciences : CMLS
Buondonno I,Gazzano E,Tavanti E,Chegaev K,Kopecka J,Fanelli M,Rolando B,Fruttero R,Gasco A,Hattinger C,Serra M,Riganti C
Fri Feb 01 00:00:00 EST 2019
Hyaluronated liposomes containing H2S-releasing doxorubicin are effective against P-glycoprotein-positive/doxorubicin-resistant osteosarcoma cells and xenografts.
Cancer letters
Gazzano E,Buondonno I,Marengo A,Rolando B,Chegaev K,Kopecka J,Saponara S,Sorge M,Hattinger CM,Gasco A,Fruttero R,Brancaccio M,Serra M,Stella B,Fattal E,Arpicco S,Riganti C
Thu Aug 01 00:00:00 EDT 2019
TRIB3 Stabilizes High TWIST1 Expression to Promote Rapid APL Progression and ATRA Resistance.
Clinical cancer research : an official journal of the American Association for Cancer Research
Lin J,Zhang W,Niu LT,Zhu YM,Weng XQ,Sheng Y,Zhu J,Xu J
Tue Oct 15 00:00:00 EDT 2019
TRIB3 Stabilizes High TWIST1 Expression to Promote Rapid APL Progression and ATRA Resistance.
Clinical cancer research : an official journal of the American Association for Cancer Research
Lin J,Zhang W,Niu LT,Zhu YM,Weng XQ,Sheng Y,Zhu J,Xu J
Tue Oct 15 00:00:00 EDT 2019
Tribbles Homolog 3 Involved in Radiation Response of Triple Negative Breast Cancer Cells by Regulating Notch1 Activation.
Cancers
Lee YC,Wang WL,Chang WC,Huang YH,Hong GC,Wang HL,Chou YH,Tseng HC,Lee HT,Li ST,Chen HL,Wu CC,Yang HF,Wang BY,Chang WW
Tue Jan 22 00:00:00 EST 2019
TRIB3 suppresses proliferation and invasion and promotes apoptosis of endometrial cancer cells by regulating the AKT signaling pathway.
OncoTargets and therapy
Qu J,Liu B,Li B,Du G,Li Y,Wang J,He L,Wan X
Fri Apr 08 00:00:00 EDT 2022
Loss of C/EBP-β LIP drives cisplatin resistance in malignant pleural mesothelioma.
Lung cancer (Amsterdam, Netherlands)
Kopecka J,Salaroglio IC,Righi L,Libener R,Orecchia S,Grosso F,Milosevic V,Ananthanarayanan P,Ricci L,Capelletto E,Pradotto M,Napoli F,Di Maio M,Novello S,Rubinstein M,Scagliotti GV,Riganti C
Fri Jun 01 00:00:00 EDT 2018
Loss of C/EBP-β LIP drives cisplatin resistance in malignant pleural mesothelioma.
Lung cancer (Amsterdam, Netherlands)
Kopecka J,Salaroglio IC,Righi L,Libener R,Orecchia S,Grosso F,Milosevic V,Ananthanarayanan P,Ricci L,Capelletto E,Pradotto M,Napoli F,Di Maio M,Novello S,Rubinstein M,Scagliotti GV,Riganti C
Fri Jun 01 00:00:00 EDT 2018
Increasing intratumor C/EBP-β LIP and nitric oxide levels overcome resistance to doxorubicin in triple negative breast cancer.
Journal of experimental & clinical cancer research : CR
Salaroglio IC,Gazzano E,Abdullrahman A,Mungo E,Castella B,Abd-Elrahman GEFA,Massaia M,Donadelli M,Rubinstein M,Riganti C,Kopecka J
Tue Nov 27 00:00:00 EST 2018
Increasing intratumor C/EBP-β LIP and nitric oxide levels overcome resistance to doxorubicin in triple negative breast cancer.
Journal of experimental & clinical cancer research : CR
Salaroglio IC,Gazzano E,Abdullrahman A,Mungo E,Castella B,Abd-Elrahman GEFA,Massaia M,Donadelli M,Rubinstein M,Riganti C,Kopecka J
Tue Nov 27 00:00:00 EST 2018
Two repeated low doses of doxorubicin are more effective than a single high dose against tumors overexpressing P-glycoprotein.
Cancer letters
Riganti C,Gazzano E,Gulino GR,Volante M,Ghigo D,Kopecka J
Fri May 01 00:00:00 EDT 2015
Two repeated low doses of doxorubicin are more effective than a single high dose against tumors overexpressing P-glycoprotein.
Cancer letters
Riganti C,Gazzano E,Gulino GR,Volante M,Ghigo D,Kopecka J
Fri May 01 00:00:00 EDT 2015
Transcriptome analysis of genes regulated by cholesterol loading in two strains of mouse macrophages associates lysosome pathway and ER stress response with atherosclerosis susceptibility.
PloS one
Berisha SZ,Hsu J,Robinet P,Smith JD
Mon Dec 30 00:00:00 EST 2013
Transcriptome analysis of genes regulated by cholesterol loading in two strains of mouse macrophages associates lysosome pathway and ER stress response with atherosclerosis susceptibility.
PloS one
Berisha SZ,Hsu J,Robinet P,Smith JD
Mon Dec 30 00:00:00 EST 2013