Search Thermo Fisher Scientific
FIGURE: 1 / 1
Recommended positive controls: MCF7.
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
TUBB4A (beta-4 tubulin) encodes a member of the beta tubulin family. Beta tubulins are one of two core protein families (alpha and beta tubulins) that heterodimerize and assemble to form microtubules. Mutations in this gene cause hypomyelinating leukodystrophy-6 and autosomal dominant torsion dystonia-4. Alternate splicing results in multiple transcript variants encoding different isoforms. A pseudogene of this gene is found on chromosome X.