Invitrogen
MA5-12159 targets Thyroid Stimulating Hormone in IHC (P) applications and shows reactivity with Human samples.
The MA5-12159 immunogen is purified human TSH-beta.
The four human glycoprotein hormones chorionic gonadotropin (CG), luteinizing hormone (LH), follicle stimulating hormone (FSH), and thyroid stimulating hormone (TSH) are dimers consisting of alpha and beta subunits that are associated noncovalently. The alpha subunits of these hormones are identical, however, their beta chains are unique and confer biological specificity. The protein encoded by this gene is the alpha subunit and belongs to the glycoprotein hormones alpha chain family.