Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: LFDNYMQQDA HEFLNYLLNT IADILQEEKK QEKQNGKLKN GNMNEPAENN KPELTWVHEI FQGTLTNETR CLNCETVSSK DEDFLDLSVD VEQN
Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 99%.
Binds to an epitope located within the peptide sequence TNETRCLNCE as determined by overlapping synthetic peptides.
Modification of cellular proteins by ubiquitin is an essential regulatory mechanism controlled by the coordinated action of multiple ubiquitin-conjugating and deubiquitinating enzymes. USP46 belongs to a large family of cysteine proteases that function as deubiquitinating enzymes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Deubiquitinating enzyme 46; Ubiquitin carboxyl-terminal hydrolase 46; Ubiquitin thioesterase 46; ubiquitin thiolesterase 46; Ubiquitin-specific-processing protease 46
Gene Aliases: USP46
UniProt ID: (Human) P62068
Entrez Gene ID: (Human) 64854
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support