Search Thermo Fisher Scientific
FIGURE: 1 / 2
Reconstitute with 100 µL of distilled water.
The antigen is homologous in rat and mouse.
This gene is a member of the vesicular amine transporter family. The encoded transmembrane protein transports acetylcholine into secretory vesicles for release into the extracellular space. Acetylcholine transport utilizes a proton gradient established by a vacuolar ATPase. This gene is located within the first intron of the choline acetyltransferase gene.