Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MA5-17608 targets VAMP/synaptobrevin in ELISA, FACS, and WB applications and shows reactivity with Human samples.
The MA5-17608 immunogen is human Synaptobrevin 1 (aa 1-118) and Synaptobrevin 2(aa 33-96).
Vesicle-associated membrane protein 2 is essential for cAMP-regulated exocytosis. VAMP (also know as synaptobrevin) is part of the synaptic vesicle docking and fusion complex and plays a central role in neuroexocytosis. Two VAMP (vesicle-associated membrane protein) isoforms are expressed in the nervous system and are differently distributed among the specialized parts of the tissue. VAMP-1 and -2 are present in all tissues tested, including kidney, adrenal gland, liver, pancreas, thyroid, heart, and smooth muscle. The two isoforms are differentially expressed in various tissues and their level may depend on differentiation. VAMP-1 is restricted to exocrine pancreas and to kidney tubular cells, whereas VAMP-2 is the predominant isoform present in Langerhans islets and in glomerular cells. Both isoforms show a patchy vesicular intracellular distribution in confocal microscopy. Evidence for the importance of neuronal VAMPproteins in the physiology of all cells is indicated.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: synaptobrevin 2; Synaptobrevin-1; Synaptobrevin-2; VAMP-1; VAMP-2; Vesicle-associated membrane protein 1; vesicle-associated membrane protein 1 (synaptobrevin 1); Vesicle-associated membrane protein 2; vesicle-associated membrane protein 2 (synaptobrevin 2)
Gene Aliases: SPAX1; SYB1; SYB2; VAMP-1; VAMP-2; VAMP1; VAMP2
UniProt ID: (Human) P23763, (Human) P63027
Entrez Gene ID: (Human) 6843, (Human) 6844
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support