Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: KLLLSQLDSH PSHSAVVNWT SYASSIEALS SGNKEFKGTV FFDEFTFVKL TGVAGNYTVC QKDLCCHLSY KMSENIPNEV YALGAFDGLH TVEGRYYLQI
Hematopoietic precursor cells migrate to the thymus, where they differentiate into mature T lymphocytes. GPI-anchored vanin-1 protein regulates the late adhesion steps of thymus homing of bone marrow precursor cells. Vanin-1 is ubiquitously expressed as a pantetheinase enzyme and catalyzes the hydrolysis of pantetheine for vitamin B5 recycling. The hydrolytic activity of vanin-1 generates the potent antioxidant cysteamine as a metabolite. As a membrane bound pantetheinase, vanin-1 provides the main source of cysteamine under normal physiological conditions. In mice, vanin-1 is expressed specifically in male Sertoli cells of the developing testis, where it aids in cell migration.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Pantetheinase; Pantetheine hydrolase; Tiff66; Vanin-1; Vanin1; vannin 1; Vascular non-inflammatory molecule 1
Gene Aliases: HDLCQ8; Tiff66; VNN1
UniProt ID: (Human) Q9UF16
Entrez Gene ID: (Human) 8876
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support