Search Thermo Fisher Scientific
PA1-10290 detects Vitellogenin from Senegalese sole (Solea senegalensis), sea bass (Dicentrarchus labrax) and seabream (Sparus aurata) samples.
PA1-10290 has been successfully used in Western blot and ELISA applications.
The PA1-10290 immunogen is native vitellogenin purified from estradiol induced plasma from male Senegalese sole fish.