Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSPRSCLRSL RLLVFAVFSA AASNWLYLAK LSSVGSISEE ETCEKLKGLI QRQVQMCKRN LEVMDSVRRG AQLAIEECQY QFRNRRWNCS TLDSLPVFGK VVTQGTREAA FVYAISSAGV AFAVTRACSS GELEKCGCDR TVHGVSPQGF QWSGCSDNIA YGVAFSQSFV DVRERSKGAS SSRALMNLHN NEAGRKAILT HMRVECKCHG VSGSCEVKTC WRAVPPFRQV GHALKEKFDG ATEVEPRRVG SSRALVPRNA QFKPHTDEDL VYLEPSPDFC EQDMRSGVLG TRGRTCNKTS KAIDGCELLC CGRGFHTAQV ELAERCSCKF HWCCFVKCRQ CQRLVELHTC R
The WNT gene family consists of structurally related genes which encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is a member of the WNT gene family, and is the first signaling molecule shown to influence the sex-determination cascade. It encodes a protein which shows 98% amino acid identity to the Wnt4 protein of mouse and rat. This gene and a nuclear receptor known to antagonize the testis-determining factor play a concerted role in both the control of female development and the prevention of testes formation. This gene and another two family members, WNT2 and WNT7B, may be associated with abnormal proliferation in breast tissue. Mutations in this gene can result in Rokitansky-Kuster-Hauser syndrome and in SERKAL syndrome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Protein Wnt-4; wingless-type MMTV integration site family member 4; wingless-type MMTV integration site family, member 4
Gene Aliases: SERKAL; UNQ426/PRO864; WNT-4; WNT4
UniProt ID: (Human) P56705
Entrez Gene ID: (Human) 54361
Molecular Function: intercellular signal molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support