Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MRLSQKWQVI NQRSLLCGED EAADENPESQ EMLEEQLVRM LTREVMDLIT VCCVSKKGAD HSSAPPADGD DEEMMATEVT PSAMAELTDL GKCLMKHEDV CTALLITAFN SLAWKDTLSC QRTTSQLCWP LLKQVLSGTL LADAVTWLFT SVLKGLQMHG QHDGCMASLV HLAFQIYEAL RPRYLEIRAV MEQIPEIQKD SLDQFDCKLL NPSLQKVADK RRKDQFKRLI AGCIGKPLGE QFRKEVHIKN LPSLFKKTKP MLETEVLDND GGGLATIFEP
Exportin-5 belongs to a large family of karyopherins that mediate the transport of proteins and other cargo between the nuclear and cytoplasmic compartments. Exportin-5 belongs to a large family of karyopherins that mediate the transport of proteins and other cargo between the nuclear and cytoplasmic compartments.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Exp5; Exportin-5; FLJ14239; FLJ32057; FLJ45606; Ran-binding protein 21
Gene Aliases: exp5; KIAA1291; RANBP21; XPO5
UniProt ID: (Human) Q9HAV4
Entrez Gene ID: (Human) 57510
Molecular Function: transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support