Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
A suggested positive control is A-20 cell lysate.
PA5-20614 can be used with blocking peptide PEP-0734.
ZBRK1 is a member of the Kruppel-associated box-zinc finger protein (KRAB-ZFP) protein family of transcriptional repressors. ZBRK1 has been shown to not only be up-regulated in senescent fibroblasts but also interacts with BRCA1, a transcriptional regulator with tumor suppressor activity. ZBRK1 binds to a specific sequence within the GADD45 gene, supporting the assembly of a nuclear complex containing BRCA1 and repressing its transcription. Recent experiments demonstrate that ZBRK1 can homo-oligomerize in vitro and in vivo through a C-terminal transcriptional repression domain (CTRD), and this oligomerization facilitates the ZBRK1-directed transcriptional repression through ZBRK1 response elements. These results suggest that higher order ZBRK1 oligomers may assemble through both protein-DNA and CTRD-dependent protein-protein interactions.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: KRAB zinc finger protein ZFQR; Zinc finger and BRCA1-interacting protein with a KRAB domain 1; Zinc finger protein 350; Zinc finger protein ZBRK1; zinc-finger protein ZBRK1
Gene Aliases: ZBRK1; ZFQR; ZNF350
UniProt ID: (Human) Q9GZX5
Entrez Gene ID: (Human) 59348
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support