The recommended shelf life for this product is 1 year from date of receipt.
ZBTB2 may be involved in transcriptional regulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Zinc finger and BTB domain-containing protein 2; ZNF909
Gene Aliases: Gm1103; KIAA1483; ZBTB2; ZNF437
UniProt ID: (Human) Q8N680
Entrez Gene ID: (Human) 57621, (Mouse) 381990
Molecular Function:
KRAB box transcription factor
transcription factor
zinc finger transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support