Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product # 7KMWCO
The protein encoded by this gene is a zinc finger protein containing a KRAB domain. Studies in mouse suggest that this protein may function as a transcriptional repressor. Mutations in this gene have been associated with transient neonatal diabetes mellitus type 1 (TNDM1).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DADB-25P22.4; Zfp-57; Zinc finger protein 57; Zinc finger protein 57 homolog; Zinc finger protein 698
Gene Aliases: bA145L22; bA145L22.2; C6orf40; G19; TNDM1; Zfp-57; ZFP57; ZNF698
UniProt ID: (Human) Q9NU63, (Mouse) Q8C6P8
Entrez Gene ID: (Human) 346171, (Mouse) 22715
Molecular Function:
KRAB box transcription factor
transcription factor
zinc finger transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support