Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
ZFP82 is a protein coding gene, and may be involved in transcriptional regulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Zfp-82; Zinc finger protein 545; Zinc finger protein 82 homolog; zinc finger protein 85
Gene Aliases: KIAA1948; ZFP82; ZNF545
UniProt ID: (Human) Q8N141
Entrez Gene ID: (Human) 284406
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support