Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MALTQVRLTF RDVAIEFSQE EWKCLDPAQR ILYRDVMLEN YWNLVSLGLC HFDMNIISML EEGKEPWTVK SCVKIARKPR TPECVKGVVT DLLRRWKHWL LLLGICCPKP HGRVSSRLRL SRSLGHFFHS AFATFMGVCD KRVGSIF
The protein encoded by this gene is a Kruppel-related zinc finger protein which is characterized by the presence of an N-terminal repressor domain, the Kruppel-associated box (KRAB). The KRAB domain is a potent repressor of transcription; thus this protein may function in transcription regulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HKr18; KRAB zinc finger protein KR18; Zinc finger protein 160; zinc finger protein 5; Zinc finger protein HZF5; Zinc finger protein Kr18
Gene Aliases: F11; HKr18; HZF5; KIAA1611; KR18; ZNF160
UniProt ID: (Human) Q9HCG1
Entrez Gene ID: (Human) 90338
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support