Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide Sequence: VEECPAEGKI PFWNFPEVCQ VDEQIERQHQ DDQDKCLLMQ VGFSDKKTII TKSARDCHEF GNILHLSTNL VASIQRPDKH ESFGNNMVDN LDLFSRSSAE NKYDNGCAKL
Zinc-finger proteins bind nucleic acids and play important roles in various cellular functions, including cell proliferation, differentiation, and apoptosis. This gene encodes a zinc finger protein, and belongs to the krueppel C2H2-type zinc-finger protein family. It may be involved in transcriptional regulation. Multiple alternatively spliced transcript variants encoding different isoforms have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Zinc finger protein 182; Zinc finger protein 21; zinc finger protein 21 (KOX 14); Zinc finger protein KOX14
Gene Aliases: HHZ150; KOX14; Zfp182; ZNF182; ZNF21
UniProt ID: (Human) P17025
Entrez Gene ID: (Human) 7569
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support