Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide Sequence: ERELDGPKQI FFGRRKDMIA EKLAPSEITE ELPSSQLMPV KKQLQGASWE LQSLRPHDED IKTTNVKSAS RQKTSLGIEL HCNVSNILHM NGSQSSTYRG
Isoform 1 and isoform 2 act as DNA-dependent transcriptional repressors.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Zinc finger and SCAN domain-containing protein 14; Zinc finger protein 396
Gene Aliases: ZNF396; ZSCAN14
UniProt ID: (Human) Q96N95
Entrez Gene ID: (Human) 252884
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support