The recommended shelf life for this product is 1 year from date of receipt.
ZNF444 has a domain structure and amino acid sequence similar to several zinc finger transcription factors, which strongly suggested that ZNF444 is a transcriptional regulator.ZNF444 has a domain structure and amino acid sequence similar to several zinc finger transcription factors, such as Kruppel-like ZNF191 (MIM 194534).[supplied by OMIM].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Endothelial zinc finger protein 2; EZF-2; Zinc finger and SCAN domain-containing protein 17; Zinc finger protein 444
Gene Aliases: EZF-2; EZF2; ZNF444; ZSCAN17
UniProt ID: (Human) Q8N0Y2
Entrez Gene ID: (Human) 55311
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support