Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 9
This gene, ZNF670, which is located on chromosome 1, is predicted to encode a zinc finger protein, currently with unknown fucntion.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Zinc finger protein 670
Gene Aliases: ZNF670
UniProt ID: (Human) Q9BS34
Entrez Gene ID: (Human) 93474
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support