Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Knockdown to ensure that the antibody binds to the antigen stated.
Immunogen sequence: PYSKGSGGGD SRKDSGSSSV SSTSSSSSSS PGDKAGFRVP SAACPPFPPH GAPVSASSSS S
Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 92%.
Binds to an epitope located within the peptide sequence PGDKAGFRVP as determined by overlapping synthetic peptides.
ZNF703 belongs to the Elbow/Noc family. ZNF703 contains 1 C2H2-type zinc finger. ZNF703 may function as a transcriptional repressor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Zinc finger elbow-related proline domain protein 1; Zinc finger protein 703
Gene Aliases: NLZ1; ZEPPO1; ZNF503L; ZNF703; ZPO1
UniProt ID: (Human) Q9H7S9
Entrez Gene ID: (Human) 80139
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support