Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
2-5 µg/mL | - |
Immunohistochemistry (IHC) |
2-5 µg/mL | - |
Product Specifications | |
---|---|
Species Reactivity |
Human, Rat |
Host/Isotype |
Sheep / IgG |
Class |
Polyclonal |
Type |
Antibody |
Immunogen |
Synthetic peptide corresponding to residues E(114) K E E V A E E A Q S G G D(127) of human gamma synuclein protein. |
Conjugate |
Unconjugated |
Form |
Lyophilized |
Concentration |
1 mg/mL |
Purification |
Affinity chromatography |
Storage buffer |
PBS |
Contains |
no preservative |
Storage conditions |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Glycerol (1:1) may be added for added stability. |
RRID |
AB_1075264 |
Reconstitute with 20 µL of distilled water.
This gene encodes a member of the synuclein family of proteins which are believed to be involved in the pathogenesis of neurodegenerative diseases. Mutations in this gene have also been associated with breast tumor development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Breast cancer-specific gene 1 protein; Gamma-synuclein; Persyn; Sensory neuron synuclein; SR; Synoretin; synuclein, gamma (breast cancer-specific protein 1)
Gene Aliases: BCSG1; PERSYN; PRSN; SNCG; SR
UniProt ID: (Human) O76070, (Rat) Q63544
Entrez Gene ID: (Human) 6623, (Rat) 64347
Molecular Function:
membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support