Search Thermo Fisher Scientific
MA1-21405 detects a-hCG in Human samples.
MA1-21405 has been successfully used in ELISA and Immunoprecipitation procedures.
MA1-21405 immunogen corresponds to Full length native protein (purified) (Human)..
Luteinizing hormone (LH, also known as lutropin) is a tropic hormone which modulates the secretory activity of other endocrine glands. It is produced in the anterior hypophysis of the pituitary gland. The glycoprotein hormone, LH, like follicle stimulating hormone and thyroid stimulating hormone, is composed of a common alpha-subunit but also a specific beta-subunit, which characterizes each of these hormones.