Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:500 | - |
Immunohistochemistry (IHC) |
1:50 | - |
Immunocytochemistry (ICC/IF) |
1:500 | - |
Product Specifications | |
---|---|
Species Reactivity |
Mouse, Rat |
Host/Isotype |
Rabbit / IgG |
Class |
Polyclonal |
Type |
Antibody |
Immunogen |
Peptide EGFAQENSKYNKTC, corresponding to amino acids 367-380 of rat mGluR5 |
Conjugate |
Unconjugated |
Form |
Lyophilized |
Concentration |
0.8 mg/mL |
Purification |
Antigen affinity chromatography |
Storage buffer |
PBS, pH 7.4, with 1% BSA |
Contains |
0.05% sodium azide |
Storage conditions |
-20°C |
RRID |
AB_2736146 |
Product is shipped at room temperature as a lyophilized powder and should be stored at -20 °C upon receipt. Reconstitution: add 50 µL of deionized water.
GRK1 is a member of the guanine nucleotide protein-coupled (G protein) receptor kinase subfamily of Serine/Threonine kinase family. GRK1 phosphorylates rhodopsin and initiates its deactivation. G protein mediated phosphorylation promotes the binding of arrestin, which removes the receptor from the G-protein and terminates the receptor signal chain. GRK1 is farnesylated to promote membrane association. Studies have shown that GRK1 has been shown to be inhibited by recoverin, a Calcium binding protein. Diseases associated with GRK1 protein dysfunction include Oguchi Disease-2 and Hereditary Night Blindness.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: A830007B11Rik; D16Ium24; D16Ium24e; G protein coupled receptor, family C, group 1, member E; G protein-coupled receptor GRM5; G protein-coupled receptor, family C, group 1, member E; Glur-5; Glur5; Glurbeta1; glutamate receptor; GPRC1A; GRM 1; GRM 5; GRM1; GRM1 alpha; GRM1A; metabotropic 1; metabotropic glutamate receptor (mGluR5); Metabotropic glutamate receptor 5; metabotropic glutamate receptor 5b; metabotropic glutamate receptor subtype 5b; metabotropic glutamate receptor type 5; MGC25101; mGlu1; mGluR; mGluR1 alpha; mGluR1A; mGluR5; mGluR5a
Gene Aliases: 6430542K11Rik; AI850523; Glu5R; Gprc1e; Grm5; Mglur5; mGluR5b
UniProt ID: (Rat) P31424, (Mouse) Q3UVX5
Entrez Gene ID: (Rat) 24418, (Mouse) 108071
Molecular Function:
G-protein coupled receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support