TacroType™ Pharmacogenetic Test
Dose Smart from the Start
TacroType: genotype-guided tacrolimus dosing
TacroType is a pharmacogenetic test designed to assess genetic variation in the CYP3A5 gene that affects how a transplant patient metabolizes tacrolimus. The test helps provide insights that transplant teams may use to inform dosing decisions from the very first dose.
The clinical challenge: the limits of conventional dosing
Tacrolimus is one of the most widely utilized immunosuppressive drugs provided to transplant recipients today. Yet, early tacrolimus management is challenged by wide, patient-specific variability in drug metabolism, making it difficult to achieve therapeutic levels quickly and safely.1-4 Conventional tacrolimus dosing overlooks significant interpatient differences in metabolism, even among individuals of the same race.5,6 Because tacrolimus has a narrow therapeutic window, acute rejection or injury can result from misdosing.² 
Conventional dosing limitations Impact on care
Relies on reactive adjustments rather than proactive management May lead to prolonged time to reach therapeutic exposure and delay stabilization of immunosuppression
Standard approaches necessitate frequent drug monitoring and clinician-led titration May increase demands on laboratory capacity, clinical staff time, and impact duration of patient stay
Does not account for interpatient variability in tacrolimus metabolism Patients may experience repeated blood draws, dose changes, and uncertainty
Dose smart from the start with TacroType
The TacroType test is a genotyping test that uses a buccal swab or blood sample to characterize CYP3A5 genotype information that healthcare providers may interpret and use to adjust tacrolimus dosing in their patients. The lab categorizes a patient as one of the following:
  • Poor metabolizer
  • Intermediate metabolizer
  • Rapid metabolizer
Our reports use interpretation standards aligned with CPIC (Clinical Pharmacogenetics Implementation Consortium) and DPWG (Dutch Pharmacogenetics Working Group) guidelines, helping ensure transplant teams receive current, evidence-based information.
TacroType provides:
Personalization
Metabolism-based guidance using patient-specific genetic information
Smarter dosing
Genetic data to inform early tacrolimus dosing decisions in transplant patients
Precision
Genetic insights move beyond conventional dosing approaches
Actionable insights
Information that may support clinical decision-making
The connection between the CYP3A5 gene and tacrolimus metabolism
Tacrolimus is predominantly metabolized by the cytochrome P450 3A (CYP3A) enzyme family, with CYP3A5 playing a key role in its oxidative metabolism in the liver and intestine.7 Genetic polymorphisms in the CYP3A5 gene result in differing levels of functional enzyme, which in turn can affect how rapidly tacrolimus is metabolized: individuals with at least one functional CYP3A5*1 allele (expressers) generally have lower dose-adjusted tacrolimus concentrations and may require higher doses to reach therapeutic trough levels compared with non-expressers (e.g., *3/*3 genotypes) who metabolize the drug more slowly.8 This genotype-associated variability partly explains the substantial inter-individual differences in tacrolimus pharmacokinetics seen in transplant recipients, and forms the basis of CYP3A5 genotype-guided dosing recommendations published by the Clinical Pharmacogenetics Implementation Consortium (CPIC).9
The benefits of genotype-guided dosing
Existing third-party studies highlight the potential value of genotype-guided tacrolimus dosing.
Faster time to target
A third-party study has suggested that genotype-guided dosing improves achievement of target tacrolimus levels compared with standard dosing.10
Potential cost savings
One decision-tree model cites a ~$17K savings in total institutional costs associated with care when using CYP3A5 genotype-guided dosing for pediatric heart transplant patients.11
Improved patient care
In one published analysis, pharmacogenomic-guided tacrolimus therapy was associated with ~ 6% fewer hospital-days and a lower incidence of adverse drug events, compared to standard dosing.12
A streamlined workflow for timely care
We understand that time is of the essence in the pre-transplant period. TacroType is designed to be non-invasive, with a timely turnaround.
  1. Simple Collection: A buccal swab or a blood sample may be collected at the bedside.
  2. Timely Turnaround: Once the sample reaches One Lambda Laboratories, results are typically released within 48 business hours.
  3. Secure Access: Reports are delivered via a HIPAA-compliant secure portal, with email notifications sent directly to the ordering clinician.
  4. EMR Integration: The concise, one-page report is designed for easy uploading into systems like Epic or Cerner.
Frequently asked questions
What is the TacroType Pharmacogenetic Test?
TacroType is a pharmacogenetic test designed to assess genetic variation in the CYP3A5 gene that affects how an organ transplant patient metabolizes tacrolimus.
Does TacroType replace conventional trough level or therapeutic drug monitoring?
No, TacroType is an adjunct to conventional dose monitoring methods. It helps provide patient-specific genotype data that may provide information regarding tacrolimus metabolism to aid in initial dosing decisions.
What is the expected turnaround time for the TacroType test?
Test results are typically provided within 48 business hours from the time the sample is received. Turnaround times may vary.
What is the sample input type for TacroType?
TacroType uses a buccal swab or a blood sample for evaluation.
What information does a patient’s TacroType report provide?
This assay detects the CYP3A5 *1, *3, *6, and *7 alleles, which may affect expression of the CYP3A5 enzyme responsible for tacrolimus metabolism. The genotype is used to predict metabolizer phenotype and guide initial tacrolimus dosing. On a report the lab categorizes a patient as one of the following:
  • Poor metabolizer
  • Intermediate metabolizer
  • Rapid metabolizer 
Results should be interpreted by a licensed clinician in conjunction with therapeutic drug monitoring (TDM) and clinical factors.
How might this information support transplant patient care?
The TacroType test results can help provide pharmacogenetic information relevant to tacrolimus metabolism. Individuals expressing CYP3A5 may require higher starting doses to achieve therapeutic drug concentrations compared with CYP3A5 non-expressers. Dosing considerations are based on CPIC and DPWG guidelines. Dose adjustments must be guided by measured tacrolimus trough levels, concomitant medications, and clinical response.
Get started with precision transplant care
Operational excellence: One Lambda Laboratories
TacroType is performed by One Lambda Laboratories, a CLIA-certified facility specializing in high-complexity transplant diagnostics. Our laboratory operates with a robust quality management framework to help ensure that the results that you receive are analytically reliable and consistent with validated performance characteristics..
  • Customer Support: Dedicated specialists are available at +1 (463) 234-6725 for onboarding and technical assistance.
  • Easy Ordering: Pre-labeled collection kits and shipping materials are provided to all registered accounts.
Get Started with Precision Transplant Care
Incorporate genotype-informed tacrolimus dosing into your institutions protocols.
  • Request a Sample Report: See how phenotype classifications are presented.
  • Schedule a Clinical Overview: Let our specialists walk your team through the validation data.
  • Order Collection Kits: Contact tdx-oll@thermofisher.com to equip your unit.
TacroType: Dose Smart from the Start.
References
  1. van Gelder T. Tacrolimus—Why pharmacokinetics matter in the clinic. Transplant Int. 2025;38:e15205. doi:10.3389/ti.2024.15205
  2. Staatz CE, Tett SE. Failure of achieving tacrolimus target blood concentration might be avoided by a wide genotyping of transplanted patients. J Pers Med. 2020;10(2):47. doi:10.3390/jpm10020047
  3. Varnell C, et al. Factors influencing tacrolimus target concentration attainment in pediatric kidney transplant recipients. Pediatr Transplant. 2023;27(4):e14393. doi:10.1111/petr.14393
  4. Stifft F, et al. Oral tacrolimus pharmacokinetics and dose–exposure variability early after thoracic organ transplantation. Pharmaceutics. 2020;12(1):72. doi:10.3390/pharmaceutics12010072
  5. Sanchez-Garcia et al. Am J Transplant. 2022;22(S3). “Tacrolimus Metabolizer Heterogeneity within Caucasian KT Recipients.”
  6. Dyer M et al. Nucleic Acids Res. 2025;53:D948–D957
  7. Staatz CE, Tett SE. Clinical pharmacokinetics and pharmacodynamics of tacrolimus in solid organ transplantation. Clin Pharmacokinet. 2004;43(10):623-653. doi:10.2165/00003088-200443100-00001
  8. Haufroid V, Mourad M, Van Kerckhove V, et al. The effect of CYP3A5 and MDR1 (ABCB1) polymorphisms on cyclosporine and tacrolimus dose requirements and trough blood levels in renal transplant recipients. Pharmacogenetics. 2004;14(3):147-154.
  9. Birdwell KA, Decker B, Barbarino JM, et al. Clinical Pharmacogenetics Implementation Consortium (CPIC) guideline for CYP3A5 genotype and tacrolimus dosing. Clin Pharmacol Ther. 2015;98(1):19-24. doi:10.1002/cpt.113
  10. Lloberas et al., 2023. PMID 37391040. EudraCT 2016-00340-34
  11. Wang et al., 2024. Pharmacogenetics and Genomics. 2025, 35: 81-86.
  12. Fragoulakis et al. Pharmacogenomics J. 2024 Mar 18;24(2):10. doi: 10.1038/s41397-024-00330-5. PMID: 38499549
For healthcare professional use only. TacroType is a laboratory developed test (LDT) performed by One Lambda Laboratories. This test has not been cleared or approved by the US FDA or CE marked in the EU. Clinical management must include therapeutic drug monitoring and comprehensive patient assessment.