Search
Bovine pulmonary artery endothelial cells labeled with probes for actin, mitochondria and the phosphorylated form of histone H3. Actin was labeled with blue-fluorescent Alexa Fluor® 350 phalloidin (Cat. no. A22281), and phosphorylated histone H3 was detected using a rabbit anti–phosphohistone H3 antibody prelabeled with the Zenon® Alexa Fluor® 555 Rabbit IgG Labeling Kit (red fluorescence, Cat. no. Z25305). Endogenous biotin associated with the mitochondria was labeled using streptavidin (Cat. no. S888) followed by a rabbit anti-streptavidin antibody prelabeled with the Zenon® Alexa Fluor® 488 Rabbit IgG Labeling Kit (green fluorescence, Cat. no. Z25302).
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›
Endothelial cells. Go ›
Mouse Anti-Alpha Tubulin Monoclonal Antibody (Cat. No. A11126) Go ›
Muntjac fibroblast labeled with probes for actin and the nucleus. Go ›
A prometaphase muntjac skin fibroblast stained with Alexa Fluor® 350 phalloidin, an anti–a-tubulin antibody and an anti–cdc6 peptide antibody. Go ›
Simultaneous detection of expression of five genes in a whole mount Drosophila embryo by fluorescence in situ hybridization (FISH) with five RNA probes Go ›
Fixed and permeabilized bovine pulmonary artery endothelial cells stained with Mouse anti-ATP Synthase Subunit IF1 Monoclonal Antibody. Go ›
Fixed and permeabilized HeLa cells stained with Alexa Fluor® 350 phalloidin and Mouse anti ATP Synthase Subunit IF1 Monoclonal Antibody to label actin filaments and the mitochondria, respectively. Go ›