Search
To make selection easier, products are organized into two general categories: pipette tips and automation tips.
Comparison Table
| Product lines | ||||
| Finntip Flex | SoftFit-L | |||
|---|---|---|---|---|
| Description | ||||
| Thermo Scientific ClipTip technology locks tips firmly in place with a light touch, and forms a seal on every channel. Helps eliminates the risk of loose or leaking tips. | The proprietary technology in Thermo Scientific ART universal fit barrier pipette tips delivers consistent and reliable protection from aerosol contamination, helping provide reproducible results, increasing efficiencies, and reducing costs associated with loss of samples in demanding applications. | Tips engineered as best fit for your Thermo Scientific Finnpipette to deliver optimal performance, precision, and accuracy. | Designed with a softer, flexible collar for an ergonomic fit with your Finnpipette to maximize performance and comfort. Reduces attachment and ejection forces to help reduce hand strain. | Designed to fit Rainin LTS style pipettes. Cylindrical collar shape and positive-stop design provides an exact, ergonomic fit. |
| Formats | ||||
96 and 384 |
96 |
96 and 384 |
96 |
96 |
| Volume range | ||||
12.5 – 1250 µl |
10 - 1250 µl |
10 µl – 10 mL |
10 µl – 10 mL |
20-1200 µl |
| Packaging options | ||||
Rack – hinged lid Reload insert Reload tower |
Rack – hinged lid Rack – lift off lid Reload inserts Reload tower |
Rack – hinged lid Reload inserts Reload tower |
Rack – hinged lid Reload inserts Reload tower |
Rack – hinged lid Rack – lift off lid Reload inserts Reload tower |
| Pipette compatibility | ||||
|
Gilson pipettes Eppendorf pipette
|
Rainin LTS |
|||
| Temperature range | ||||
| Frozen samples to Vapor Phase Liquid Nitrogen** | Frozen samples to Vapor Phase Liquid Nitrogen* | Ambient to -80°C* | ||
Comparison Table
| Brand | ||||
Tecan |
Beckman |
Revvity |
Qiagen | Eppendorf |
|---|---|---|---|---|
| Formats | ||||
96 |
96 |
96 |
96 |
96 |
| Tip size | ||||
10 – 1000 µl |
10-200 µl |
10-1000 µl |
300-1000 µl |
50-1000 µl |
| Liquid handler compatibility | ||||
Te-Mo Freedom EVO Genesis Fluent*
|
Biomek FX Biomek NX Biomek 3000 |
JANUS Zephyr Fontus |
BioRobot 3000 BioRobot 8000 BioRobot 9600 BioRobot 9604 |
epMotion |
For Laboratory Use. It is the customer’s responsibility to ensure that the performance of the product is suitable for customers’ specific uses or applications.