26831showcase
Fast and simple
library preparation
The AB Library Builder™ System simplifies your next-generation sequencing workflow by automating the most labor-intensive parts of library preparation for the Ion Proton™ and Ion PGM™ sequencing systems.
Next-generation sequencing applications increasingly demand scalable, automated sample preparation techniques for creating DNA and RNA libraries. With just minutes of hands-on time and high recovery rates, the AB Library Builder™ System increases throughput for a wide variety of applications including exome, whole-transcriptome, targeted resequencing, and whole-genome sequencing.
Download the specification sheet
Automated
  • Enables reduced hands-on time by more than 50% compared to the manual workflow 
  • Multiplexing for up to 96 barcoded DNA fragment libraries
Scalable
  • Prepares up to 13 libraries per run, up to 39 libraries per day 
  • Process up to 13 DNA or RNA libraries in a single run
Integrated
  • Plug-and-play reagents and push-button protocol 
  • Integrated nucleic acid purification instrument
1 Library automation
ion-torrent.par.20114.image.175.139.1.
2 Template automation
ion-torrent.par.36772.image.175.140.1.
3 Ion semiconductor sequencers
ion-torrent.par.56737.image.175.140.1.