Search
Having difficulties with your experiment?
We are dedicated to your success. Get back on track. View our expert recommendations for commonly encountered problem scenarios.
View the relevant questions below:
Beginning your experiment?
Visit our
If the media formulation contains:
*There are some exceptions. Gibco™ DMEM has always been made according to Dulbecco’s original published formulation, with 3.7g/L sodium bicarbonate. Customers have been using this medium in CO2 incubators ranging from 5–10% CO2 for decades, usually with no trouble maintaining physiological pH; this also depends on the cell type. Once cells are growing, the pH will drop (due to metabolic accumulation of lactic acid).
There are a number of events that can contribute to this:
No. Suspension cultures will quickly become oxygen limited and will only reach cell densities of 1 to 1.5 x 10E6 cells/mL. Recombinant product expression will also be greatly reduced. We recommend using plastic Erlenmeyer shake flasks with screw cap tops (not foam plugs). This allows you to cover and keep the cultures sterile and allow free gas exchange.
Humidification is only necessary for monolayer cultures and suspension cultures with volumes less than 35 mL. With shaker or spinner cultures greater than 35 mL, humidification is not necessary.
Please see the possible causes and solutions we recommend below:
Reason | Solution |
Cells were stored incorrectly | Obtain a new stock and store in liquid nitrogen. Keep cells in liquid nitrogen until thawing. |
Homemade freezer stock is not viable | Freeze cells at a density recommended by the supplier. Use low-passage cells to make your own freezer stocks. Follow procedures for freezing cells exactly as recommended by the supplier. Obtain a new stock. |
Cells were thawed incorrectly | Follow procedures for thawing cells exactly as recommended by the supplier. Make sure that you thaw the frozen cells quickly but dilute them slowly using pre-warmed growth medium before plating. |
Thawing medium is not correct | Use the medium recommended by the supplier. Make sure the medium is pre-warmed. |
Cells are too dilute | Plate thawed cells at the highest density recommended by the supplier to optimize recovery. |
Cells not handled gently | Freezing and thawing procedures are stressful to most cells. Do not vortex, bang the flasks to dislodge the cells (except when culturing insect cells), or centrifuge cells at high speeds. |
Glycerol used in the freezing medium was stored in light (if applicable) | If stored in light, glycerol gets converted into acrolein, which is toxic to cells. Obtain a new stock. |
Please see some common reasons below, and solutions to the issue.
Reason | Solution |
Growth medium is not correct | Use pre-warmed growth medium as recommended by the supplier. |
Serum in the growth medium is of poor quality | Use serum from a different lot. |
Cells have been passaged too many times | Use healthy, low–passage number cells. |
Cells were allowed to grow beyond confluency | Passage mammalian cells when they are in the log-phase before they reach confluency. |
Culture is contaminated with mycoplasma | Discard cells, media, and reagents. Obtain new stocks of cells, and use them with fresh media and reagents. |
Please see the possible causes for this rapid pH shift, and suggested solutions:
Possible cause | Suggested solution |
Incorrect carbon dioxide tension | Increase or decrease percentage of carbon dioxide in the incubator based on concentration of sodium bicarbonate in the medium. For sodium bicarbonate concentrations of 2.0 to 3.7 g/L, use carbon dioxide amounts of 5–10%, respectively. Switch to carbon dioxide–independent medium. |
Overly tight caps on tissue culture flasks | Loosen caps one-quarter turn. |
Insufficient bicarbonate buffering | Add HEPES buffer to a final concentration of 10–25 mM. |
Incorrect salts in medium | Use an Earle’s salts-based medium in a CO2 environment and a Hanks’ salts-based medium in atmospheric conditions. |
Bacterial, yeast, or fungal contamination | Discard culture and medium. Try to decontaminate culture. |
The precipitate could be due to residual phosphate left over from detergent washing, which may precipitate powered medium components. Rinse glassware in deionized, distilled water several times, then sterilize. If the medium is frozen, try warming media to 37°C and swirl to dissolve. If precipitate remains, discard medium.
This is most likely a sign of bacterial or fungal contamination. We would recommend discarding the media, and trying to decontaminate the culture.
This can occur if cells are overly trypsinized. Trypsinize for a shorter time or use less trypsin. Mycoplasma contamination could also cause this problem. Segregate your culture and test for mycoplasma infection. Lastly, check for attachment factors in the medium.
Try changing the medium or serum. Compare media formulations for differences in glucose, amino acids, and other components. Compare an old lot of serum with a new lot. Increase initial cell inoculums. Lastly, adapt cells sequentially to new medium.
Very often mycoplasma contamination cannot be removed from the culture so it should be discarded. You may have a unique culture that you prefer not to discard and would like to try to clean it. Ciprofloxacin and Plasmocin have reportedly been used for this application. If interested in a protocol or directions for use, check with the antibiotic supplier or published literature. Note that mycoplasma are very difficult to remove from culture and spread easily so the treated cultures should be quarantined until clear of mycoplasma, and your laboratory should be thoroughly cleaned.
Decontamination of cultures with antibiotics
When an irreplaceable culture becomes contaminated, researchers may attempt to eliminate or control the contamination.
Antibiotics and antimycotics at high concentrations can be toxic to some cell lines. Therefore, perform a dose-response test to determine the level at which an antibiotic or antimycotic becomes toxic. This is particularly important when using an antimycotic such as Gibco™ Fungizone™ reagent or an antibiotic such as tylosin.
The following is a suggested procedure for determining toxicity levels and decontaminating cultures:
When L-glutamine is in a concentrated stock solution it easily precipitates when cooled. Warming the solution briefly in a 37°C water bath with gentle swirling will dissolve the precipitate. Do not use the product unless the precipitate is fully dissolved.
In all media containing GlutaMAX™ supplement dipeptides as a substitute for L-glutamine, concentration is equimolar with the L-glutamine in the original formulation.
Yes. If you suspect that this is the case, remove the medium and add fresh medium. Alternatively, you can supplement medium with growth-promoting components. It is also possible to substitute GlutaMax™ I or II for glutamine in the medium to prevent glutamine exhaustion.
We routinely ship media that require long-term storage in the refrigerator at room temperature. We have done studies on representative media formulations to show that media can be at room temperature for up to a week without a problem.
Generally speaking, media can be used for up to three weeks after supplementation with serum. There are no formal studies to support this, but it is the rule of thumb used by our scientists.
Liquid media made from dry powder media should be stable for as long as the same formulation sold in liquid format. This is usually 1 month provided it is stored refrigerated and protected from light.
Gibco™ FBS is not pre-aged. When stored at 2 to 8°C, the possibility exists for various proteins and lipoproteins in serum (e.g., cold agglutinins, fibrinogen, vitronectin, etc.) to aggregate, and form either perceptible material or observed turbidity. This should not affect serum performance. We recommend that you store FBS at –20°C and avoid repeated freeze-thaw cycles.
Flocculence may appear in FBS for a variety of reasons. The most common reason is the denaturation of serum lipoproteins. You may observe fibrin, one of the clot-forming proteins present in serum, after the serum has been thawed. This should not affect product performance.
To remove the flocculence, transfer the serum to sterile tubes and centrifuge the material briefly at 400 x g. Then filter the resulting supernant along with your media. Do not attempt to filter serum containing flocculence, it may clog filters.
Our studies have shown that short-term storage of thawed FBS at 4°C for up to 28 days causes no decrease in growth or viability performance. FBS stored at 4°C for longer periods of time should be evaluated on a case-by-case basis should this storage condition be necessary.
CD FortiCHO™ medium has proven to be an excellent single-stream medium for CHO production, clone development, and banking. In our experience, CHO clones created and selected in CD FortiCHO™ medium can be put directly into Dynamis™ medium without any adaptation. In fact, our work suggests that those cells will produce as well or better in Dynamis™ medium and are just as stable in Dynamis™ medium if used for process development and scale-up.
24.8 grams of Dynamis™ Medium powder is required for 1 liter of media.
Please refer to the sequential adaptation procedure shown below. If you have trouble getting past a certain ratio, passage the cells 2–3 more times at the previous ratio.
Generally speaking, trypsin can be stored refrigerated for up to a week. However, this may vary depending on how often it is warmed up and used during that week. It may also depend on how tightly the cells are attached, because there will be some loss of activity each day the trypsin is not frozen.
Trypsinize this type of cells for a shorter duration or use less trypsin or Gibco™ TrypLE™ Select or Gibco™ TrypLE™ Express Trypsin Replacement. After trypsinization dilute in 2 to 5 mL of cell culture growth medium transfer the cell suspension to 15 mL centrifuge tube. Centrifuge for 5–10 minutes at 100 x g. Discard the supernatant and suspend cell pellet in 2–5 mL fresh medium. Determine the count and seed the flask according to normal protocol and cell should adhere to the surface.
For Research Use Only. Not for use in diagnostic procedures.