Search
Accomplish the most challenging analytical tasks in research and manufacturing with powerful X-ray fluorescence technology for nondestructive elemental analysis of any sample. The Thermo Scientific ARL QUANT’X EDXRF spectrometer is a convenient and cost-effective analysis tool delivering major, minor and trace element quantification from F (C*) to Am for a wide range of samples with minimal sample preparation. The ARL QUANT’X offers top performance from light elements to heavy elements, speed and flexibility in a compact benchtop format.
Request a demo to speak with a member from our team and learn more about the ARL QUANT’X EDXRF Spectrometer.
* With optional SDD500G detector
The ARL QUANT’X EDXRF spectrometer provides several advantages:
Unattended instrument operation enables users to maximize sample throughput and frees up time to focus on other tasks. Our 10 and 20 position auto-changers are designed for batch analysis, which is easily started for an entire tray of samples in the software interface. Share analysis data without delay to a LIMS using the optional TraceCom package, enabling faster decision making in QC and production environments.
New compounds, one-off samples and materials without suitable reference materials are challenging to analyze. With the UniQuant advanced standardless analysis you can obtain reliable elemental composition results within minutes for any sample, including multi-element solids, pressed powder, small samples, liquids or air-filters. UniQuant comes pre-calibrated and ready to use, so everyone can get high-quality results from day one.
Accuracy and precision are the foundation for elemental analysis results you can trust. The ARL QUANT’X uses an optimized direct excitation geometry to bring top performance to your lab. Benefit from better sensitivity across the periodic table with the combination of a high power 50W X-ray tube and the latest generation SDD silicon drift detector, for both bulk and small samples.
ARL QUANT’X spectrometers are backed by our global applications and support team. With dedicated support for most countries and regions around the world, our customer support organization is the best in the industry. We offer a full suite of product and customer services tailored to bulk elemental analysis. Let us recommend a support plan or ask us how we can customize services specifically to your requirements.
The ARL QUANT’X EDXRF spectrometer is sold as a configured system designed to meet your specific needs.
| ARL QUANT’X configuration | ||||
Specification |
Select |
Ideal |
Enhance |
|
Elemental range |
Mg – U |
F - U |
C - U |
|
Atmosphere |
Air |
Air Vacuum or He |
Air Vacuum or He |
|
Detector |
SDD500 |
SDD500 |
SDD500G |
|
Sample chamber |
Large chamber with single position stage |
|||
Additional functionality |
Optional accessory |
|||
Liquids analysis |
He atmosphere |
|||
Small spot analysis |
Choice of collimators |
|||
Sample viewing |
CCD camera |
|||
Sample changers |
10 or 20 pos. sample changer |
|||
XL samples |
Chamber extension, for samples heights up to 36cm |
|||
Standardless analysis |
UniQuant software |
|||
Data transfer |
TRACEcom for interfacing with LIMS |
|||
The ARL QUANT’X EDXRF Spectrometer delivers reliable elemental analysis results in central and contract laboratories, academic institutions as well as the environmental monitoring, RoHS and WEEE screening, chemicals, mining, forensics, food, cement, and metals industries.
Read below to learn more about selected applications or contact us to find out more
Contact us today for a quote, demo, or to speak to a specialist about your laboratory XRF needs.