Search
Immunofluorescent detection of pan cadherin in MDCK cells. Confluent monolayers were fixed in 50%methanol/50%Acetone, blocked for at least 30 minutes in 1% BSA then incubated 2 hours with a pan cadherin antibody (Product # 71-7100) at 2.5 ug/mL, washed, then incubated 1 hour with Alexa Flour 594 conjugated Donkey anti-Rabbit secondary antibody (Product # A21207) at 1:2000 dilution. Cells were counterstained with DAPI (blue). Coverslips were mounted with Prolong Gold Antifade reagent (Product # P36930) and imaged at 40X. Images generated by Joell Solan in Paul Lampe Lab at the Fred Hutchinson cancer Research Center.