Search
Our techs are here to answer your questions, just email us.
If you like this month's Q&A, check out our Talk Tech archive.
Moisture in high humidity areas has been found to accumulate in the tubing attached to the pressure sensor board. Eventually this moisture migrates to the sensors and causes them to fail as outlined in the alarms/symptoms section listed below. There can be other causes for the sensor board to fail such as moisture accumulating on the actual board or just a faulty sensor but the alarms/symptoms will generally always be the same.
Alarms/Symptoms
Many changes have been made to the sensor boards to improve and protect against moisture damage.
If you have additional questions, call the Technical Support team at (866) 282-0430 and select option 2.
Talk Tech is published here every month. Receive our monthly Talk Tech answers delivered right to your inbox by subscribing using the button below.