Filters are commonplace in today’s society, and fulfill critical functions in industry and the household, ranging from air filters that give you clean, breathable air to water filters that ensure you have safe, potable tap water.
The key to high-performance filtration is the development of synthetic fibers. To optimize a filter’s design (so it continues to filter a broad range of dry or wet particulates over time), features like fiber diameter, density, and surface morphology all need to be thoroughly characterized and optimized.
Traditional methods for fiber analysis, such as optical microscopy and atomic force microscopy (AFM), are insufficient for this multi-modal analysis. Optical microscopy lacks the resolution necessary to accurately study nanofibers, whereas AFM is too slow and suffers from physical probe issues. These methods also rely on slow, manual measurement and data analysis.
Scanning electron microscopy (SEM), meanwhile, offers exceptional resolution capabilities, and allows you to automatically characterize multiple fibers at the same time. With the addition of leading automation software, critical filter parameters can be extracted rapidly and reliably. This minimizes the time spent using SEM in the lab, increasing development throughput as you can quickly and easily analyze fibers for filtration design.
Thermo Fisher Scientific offers a range of SEM instrumentation for fiber analysis. When combined with Thermo Scientific FiberMetric Software, accurate, automated measurement of micro- and nanofiber properties is possible.
Electrospun imaged with Phenom Desktop SEM.
Electrospun nanofibers on a paper substrate, imaged with a Phenom Desktop SEM.
Dust filter imaged with Phenom Desktop SEM.
Fibers from a dust filter imaged with a Phenom Desktop SEM.
Resources
Applications
Process Control_Thumb_274x180_144DPI
Controle de processo
A indústria moderna exige alta produtividade com qualidade superior, um equilíbrio mantido por meio de um controle de processo robusto. As ferramentas SEM e TEM com software de automação dedicado proporcionam informações rápidas e em várias escalas para monitoramento e aprimoramento de processos.
Quality Control_Thumb_274x180_144DPI
Controle de qualidade
O controle de qualidade e a garantia de qualidade são essenciais na indústria moderna. Oferecemos uma gama de ferramentas de microscopia eletrônica e espectroscopia para análises multidimensionais e multimodais de defeitos, permitindo que você tome decisões confiáveis e informadas para controle e melhoria de processos.
Fundamental Materials Research_R&D_Thumb_274x180_144DPI
Pesquisa de materiais fundamentais
Novos materiais são investigados em escalas cada vez menores para o máximo controle de suas propriedades físicas e químicas. A microscopia eletrônica fornece aos pesquisadores percepções importantes sobre uma ampla variedade de características materiais em escala micro a nano.
Style Sheet for Komodo Tabs
Techniques
Caracterização de materiais 3D
O desenvolvimento de materiais muitas vezes requer caracterização 3D em várias escalas. Os instrumentos DualBeam permitem a secção em série de grandes volumes e a subsequente geração de imagens SEM nanométricas, que podem ser processadas em reconstruções 3D de alta qualidade da amostra.
Saiba mais ›
Análise elementar EDS
A EDS proporciona informações de composição vitais para observações em microscópio eletrônico. Principalmente, nossos exclusivos sistemas detectores Super-X e Dual-X adicionam opções para maior produtividade e/ou sensibilidade, permitindo otimizar a aquisição de dados para atender às prioridades de pesquisa.
Saiba mais ›
ColorSEM
Usando a EDS (espectroscopia de raios X por energia dispersiva) com quantificação ao vivo, a tecnologia ColorSEM transforma as imagens SEM em uma técnica colorida. Agora, qualquer usuário pode adquirir dados elementares continuamente para obter informações mais completas do que nunca.
Saiba mais ›
Corte transversal
O corte transversal fornece dados adicionais ao revelar informações de subsuperfície. Os instrumentos DualBeam apresentam excelentes colunas de feixe de íons focalizados para corte transversal de alta qualidade. A automação permite realizar o processamento autônomo de amostras com alta produtividade.
Saiba mais ›
Caracterização de materiais 3D
O desenvolvimento de materiais muitas vezes requer caracterização 3D em várias escalas. Os instrumentos DualBeam permitem a secção em série de grandes volumes e a subsequente geração de imagens SEM nanométricas, que podem ser processadas em reconstruções 3D de alta qualidade da amostra.
Saiba mais ›
Análise elementar EDS
A EDS proporciona informações de composição vitais para observações em microscópio eletrônico. Principalmente, nossos exclusivos sistemas detectores Super-X e Dual-X adicionam opções para maior produtividade e/ou sensibilidade, permitindo otimizar a aquisição de dados para atender às prioridades de pesquisa.
Saiba mais ›
ColorSEM
Usando a EDS (espectroscopia de raios X por energia dispersiva) com quantificação ao vivo, a tecnologia ColorSEM transforma as imagens SEM em uma técnica colorida. Agora, qualquer usuário pode adquirir dados elementares continuamente para obter informações mais completas do que nunca.
Saiba mais ›
Corte transversal
O corte transversal fornece dados adicionais ao revelar informações de subsuperfície. Os instrumentos DualBeam apresentam excelentes colunas de feixe de íons focalizados para corte transversal de alta qualidade. A automação permite realizar o processamento autônomo de amostras com alta produtividade.
Saiba mais ›
Products
Folha de estilo para cartões de instrumentos original
Phenom Pharos Desktop SEM
  • Fonte FEG com faixa de tensão de aceleração de 2 kV a 15 kV
  • Resolução de <2,5 nm (SE) e <4,0 nm (BSE) a 15 kV; ampliação de até 1.000.000 vezes
  • Detector EDS e SE opcional totalmente integrado
Phenom XL G2 Desktop SEM
  • Para amostras grandes (100 x 100 mm) e ideal para automação
  • Resolução <10 nm e ampliação de até 200.000 vezes; tensão de aceleração de 4,8 kV até 20 kV
  • Detector EDS e BSE opcional totalmente integrado
Phenom Pro Desktop SEM
  • SEM de mesa de alto desempenho
  • Resolução <8 nm (SE) e <10 nm (BSE); ampliação de até 150.000 vezes
  • Detector SE opcional
Phenom ProX Desktop SEM
  • SEM de mesa de alto desempenho com detector EDS integrado
  • Resolução <8 nm (SE) e <10 nm (BSE); ampliação de até 150.000 vezes
  • Detector SE opcional
Phenom Pure Desktop SEM
  • SEM de mesa de nível básico
  • Resolução <25 nm; ampliação de até 65.000 vezes
  • Fonte CeB6 de longa duração
Phenom ParticleX AM Desktop SEM
  • SEM de mesa versátil com software de automação para fabricação aditiva
  • Resolução <10 nm; ampliação de até 200.000 vezes
  • Detector SE opcional
Phenom ParticleX TC Desktop SEM
  • Desktop SEM versátil com software de automação para limpeza técnica
  • Resolução <10 nm; ampliação de até 200.000 vezes
  • Detector SE opcional
Phenom Perception GSR Desktop SEM
  • GSR de mesa SEM automatizado dedicado
  • Resolução <10 nm; ampliação de até 200.000 vezes
  • Fonte CeB6 de longa duração
Nexsa XPS
  • Módulo de inclinação para medições ARXPS
  • Fonte de íons de modo duplo para recursos de perfil de profundidade expandida
  • Análise de isolantes
K-Alpha XPS
  • Espectroscopia de área selecionável
  • Monocromador microfocalizado
  • Espectroscopia de estado químico de alta resolução
ESCALAB Xi+ XPS
  • Espectroscopia de alta sensibilidade
  • XPS com raios X não monocromáticos
  • Analisador de energia hemisférica de 180 graus
Style Sheet to change H2 style to p with em-h2-header class
Contact us