Search
We are dedicated to your success. View our expert recommendations for commonly encountered inquiries.
View the relevant questions below:
Beginning your experiment? Visit
You may clean the source housing, sweep cone, ion transfer tube, and the RF lens behind the ITT.
If you notice poor sensitivity or the presence of large background ions, your instrument is in need of cleaning.
Need more information? Contact us ›
For Research Use Only. Not for use in diagnostic procedures.