Search
| Components | Molecular Weight | Concentration (mg/L) | mM |
|---|---|---|---|
| Inorganic Salts | |||
| Calcium Chloride (CaCl2) (anhyd.) | 111.0 | 100.0 | 0.9009009 |
| Magnesium Chloride (MgCl2-6H2O) | 203.0 | 100.0 | 0.49261084 |
| Potassium Chloride (KCl) | 75.0 | 200.0 | 2.6666667 |
| Potassium Phosphate monobasic (KH2PO4) | 142.0 | 20.0 | 0.14084508 |
| Sodium Chloride (NaCl) | 58.0 | 8000.0 | 137.93103 |
| Sodium Phosphate dibasic (Na2HPO4-7H2O) | 268.0 | 2160.0 | 8.059702 |
| Other Components | |||
| Colcemid(TM) | 371.0 | 10.0 | 0.026954178 |