Advanced features for precision temperature control 
TSC Series Recirculating Chillers are engineered to deliver precise and reliable temperature control across a wide range of applications, from laboratory experiments to industrial processes. Designed with user-centric features, these chillers ensure optimal performance and efficiency.
Flexible configurations to fit your unique space
With a broad range of cooling capacities, extended temperature set points, and flexible condenser options, you can select a configuration that meets your operational needs while maximizing efficiency.
Wide temperature range and cooling capacity
TSC Series Recirculating Chillers deliver precise temperature control from -5°C to +90°C with active heating for fast ramping, ensuring processes reach target conditions quickly and maintain stable, consistent performance. With seven cooling capacity options ranging from 0.8 kW to 26 kW, these chillers support applications from small laboratory experiments to large production-scale processes, providing the flexibility to meet diverse operational requirements.
Cooling capacity curves
T2 pump curves
C1 & C2 pump curves
Air or water-cooled condenser  
The water-cooled condenser reduces heat rejection into the surrounding environment by utilizing facility water, keeping workspaces cooler and more comfortable. This is particularly advantageous for high-capacity systems or areas sensitive to added heat, enhancing overall operational efficiency.
The air-cooled condenser simplifies installation by eliminating the need for facility water connections. It provides reliable cooling performance, making it ideal for laboratories and industrial sites where water supply may be limited.
Durable and space-efficient
Constructed with high-quality, robust materials, these chillers are designed to withstand demanding environments and continuous use. Despite their durability, they maintain a compact footprint, making them suitable for space-constrained lab or industrial setups.
Global voltage
Automatic frequency detection allows the chiller to operate seamlessly anywhere in the world. This ensures consistent performance across varying power standards, supporting international laboratories and manufacturing facilities.
Superior temperature stability and uniformity
Advanced microprocessor controls, flow and pressure monitoring, and digital communication capabilities provide reliable, outstanding performance. Whether you’re running delicate laboratory experiments or high-demand industrial processes, the TSC Series Recirculating Chillers ensures consistent results you can trust.
Flow and pressure control with real-time readout
TSC Series Recirculating Chillers provide precise fluid flow adjustment and pressure relief, ensuring optimal temperature control and protecting your system from overpressure. The integrated real-time flow and pressure readouts allow continuous monitoring, helping operators maintain consistent, reliable performance while safeguarding equipment and enabling stable, repeatable results- critical for sensitive laboratory and industrial applications.
Multiple pump options
TSC Series Recirculating Chillers offer up to four pump options per capacity step, allowing you to select the right configuration for your specific flow and pressure requirements. Choose from centrifugal or turbine pumps to match your application needs, ensuring optimal performance and flexibility across laboratory and industrial processes.
Seamless connectivity and remote control
These chillers offer comprehensive digital and analog communication options for full remote operation, monitoring, and data logging. Operators can adjust setpoints, track performance, and access historical data either via a connected system or through Thermo Cloud using the OPC-UA™ standard, enabling seamless integration into automated laboratory or industrial workflows, enhancing process control, and simplifying reporting.
OPC-UA is a trademark of OPC Foundation.
Simplifying operation through innovation
Simplified operation and intuitive controls make these chillers effortless to use.
Updated controller and user-friendly interface
LCD multi-line alphanumeric display with intuitive controls allows easy monitoring and adjustment of temperatures and parameters.
Outstanding performance meets sustainability
TSC Series Recirculating Chillers with its energy-efficient, environmentally responsible design, helps you achieve both operational excellence and sustainability goals.
Significant reduction in global warming potential (GWP) with low-GWP refrigerants
The TSC Series drastically lowers the environmental impact of cooling processes compared to conventional models. By incorporating environmentally responsible, low-GWP refrigerants, these chillers maintain precise temperature control while helping laboratories and industrial facilities meet sustainability goals and support global efforts to combat climate change- all without compromising high performance.
Energy-efficient design
Optimized engineering maximizes energy efficiency through high-performance compressors, advanced refrigeration cycles, and intelligent controls. This reduces electricity and water usage, lowers operating costs, and minimizes heat rejection into the workspace.
Compliance and cleanroom compatibility
Meets CE, RoHS, SEMI, GMP, and ISO standards. Ensures cleanroom compatibility and supports sensitive industrial or laboratory processes. Compliance helps ensure safe, reliable operation while minimizing environmental and operational risk.
Application versatility
The TSC Series Recirculating Chillers are engineered to support a wide range of laboratory and industrial processes, from small bench-top experiments to large-scale production. With precise temperature control, flexible cooling capacities, and a robust design, these chillers are ideal for: 
  • Analytical and research labs – Ensure accurate and stable temperatures for sensitive experiments.
  • Biotech apnd pharmaceutical rocesses – Support reproducible conditions for drug development, testing, and production.
  • Industrial cooling – Provide reliable cooling for machinery and manufacturing processes.
  • Laser and metrology applications – Maintain consistent thermal management for precision equipment.
  • Packaging and printing – Ensure temperature stability for high-quality production outcomes.
  • Battery testing and manufacturing – Deliver precise temperature control for safe, reliable battery testing and production processes.
  • Semi-conductors
These chillers empower you to achieve reliable, high-performance temperature control today and embrace the future of your processes tomorrow. 
For Laboratory Use. It is the customer’s responsibility to ensure that the performance of the product is suitable for customers’ specific uses or applications.